2,5-Diamino-4,6-dichloropyrimidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4,6-Dichloropyrimidine-2,5-diamine
D4231-1G
D4231-5G
TCD4231-1GEA
47.4
CAD
TCD4231-1G
TCD4231-5G
2,5-Diamino-4,6-dichloropyrimidine ≥98.0% (by GC, titration analysis)
2,5-Diamino-4,6-dichloropyrimidine
CAS Number: 55583-59-0
MDL Number: MFCD00792652
Molecular Formula: C4H4Cl2N4
Molecular Weight: 179.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181
Formula:
C₄H₄Cl₂N₄ MW: 179.0100 g/mol Melting Pt: 181 °C |
MDL Number:
MFCD00792652 CAS Number: 55583-59-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...