1,8-Dibromoanthracene ≥98.0%

Supplier: TCI America

D4230-1G D4230-5G
TCD4230-1GEA 248.06 CAD
TCD4230-1G TCD4230-5G
1,8-Dibromoanthracene ≥98.0%
1,8-Dibromoanthracene

CAS Number: 131276-24-9
Molecular Formula: C14H8Br2
Molecular Weight: 336.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168

Formula: C₁₄H₈Br₂
MW: 336.03 g/mol
Melting Pt: 168 °C
CAS Number: 131276-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01c4548cf23b28f9b33669d3afcb20f3fb2affa6&vid=01c4548cf23b28f9b33669d3afcb20f3fb2affa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR