2,5-Dibromopyrimidine ≥98.0% (by GC)

Supplier: TCI America

D4225-1G D4225-5G
TCD4225-1GEA 33.18 CAD
TCD4225-1G TCD4225-5G
2,5-Dibromopyrimidine ≥98.0% (by GC)
2,5-Dibromopyrimidine

CAS Number: 32779-37-6
MDL Number: MFCD08275684
Molecular Formula: C4H2Br2N2
Molecular Weight: 237.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85

Formula: C₄H₂Br₂N₂
Melting Pt: 85 °C
MDL Number: MFCD08275684
CAS Number: 32779-37-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf5f745c274153870cfea560b03461be2dfa91be&vid=cf5f745c274153870cfea560b03461be2dfa91be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR