2,2'-Dibromo-1,1'-binaphthyl ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
racemic-2,2'-Dibromo-1,1'-binaphthyl, 2,2-Dibromo-1,1-binaphthyl, 2,2'-Dibromo-1,1'-binaphthalene
D4221-1G
D4221-5G
TCD4221-1GEA
120.51
CAD
TCD4221-1G
TCD4221-5G
2,2'-Dibromo-1,1'-binaphthyl ≥96.0% (by GC)
2,2'-Dibromo-1,1'-binaphthyl
CAS Number: 74866-28-7
MDL Number: MFCD00188005
Molecular Formula: C20H12Br2
Molecular Weight: 412.12
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 184
Formula:
C₂₀H₁₂Br₂ MW: 412.12 g/mol Melting Pt: 184 °C |
MDL Number:
MFCD00188005 CAS Number: 74866-28-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...