2'-Deoxy-5-(hydroxymethyl)cytidine ≥98.0%
Supplier: TCI America
Synonyms:
5-hmdC
D4220-200MG
D4220-50MG
TCD4220-200MGEA
1006.46
CAD
TCD4220-200MG
TCD4220-50MG
2'-Deoxy-5-(hydroxymethyl)cytidine ≥98.0%
2'-Deoxy-5-(hydroxymethyl)cytidine
CAS Number: 7226-77-9
MDL Number: MFCD09842113
Molecular Formula: C10H15N3O5
Molecular Weight: 257.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 205
Lambda max.: 274 nm (H2O)
Product Categories: Bioscience, Nucleosides and Their Analogs, Methylated Nucleosides
Other Product Names / Synonyms: 5-hmdC
Formula:
C₁₀H₁₅N₃O₅ MW: 257.25 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD09842113 CAS Number: 7226-77-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...