Di-tert-butyl-N,N-diisopropylphosphoramidite ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Di-tert-butoxy(diisopropylamino)phosphine, Di-tert-butyl N,N-diisopropylphosphoramidite, Di-tert-butyl diisopropylphosphoramidite

D4211-1G D4211-5G
TCD4211-1GEA 71.1 CAD
TCD4211-1G TCD4211-5G
Di-tert-butyl-N,N-diisopropylphosphoramidite ≥98.0% (by total nitrogen basis)
Di-tert-butyl-N,N-diisopropylphosphoramidite

CAS Number: 137348-86-8
MDL Number: MFCD00153506
Molecular Formula: C14H32NO2P
Molecular Weight: 277.39
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 113
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Di-tert-butoxy(diisopropylamino)phosphine

Formula: C₁₄H₃₂NO₂P
MW: 277.39 g/mol
Boiling Pt: 77 °C
Density: 0.89 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00153506
CAS Number: 137348-86-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR