(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane ≥98.0% (by GC)

Supplier: TCI America

D4210-1G D4210-5G
TCD4210-1GEA 57.83 CAD
TCD4210-1G TCD4210-5G
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane ≥98.0% (by GC)
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane

CAS Number: 151213-40-0
MDL Number: MFCD08458306
Molecular Formula: C7H14N2
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 91
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: -7 deg (C=0.5, CHCl3)
Storage Temperature: <0°C


Other Product Names / Synonyms: (4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine

Formula: C₇H₁₄N₂
Boiling Pt: 59 °C
Density: 1.03 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD08458306
CAS Number: 151213-40-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88e9aa45429c07a0a0cdb6ae316682f4ad30e72e&vid=88e9aa45429c07a0a0cdb6ae316682f4ad30e72e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR