3-(1,3-Dithian-2-ylidene)-2,4-pentanedione ≥98.0% (by GC)

Supplier: TCI America

D4208-1G D4208-5G
TCD4208-1GEA 153.26 CAD
TCD4208-1G TCD4208-5G
3-(1,3-Dithian-2-ylidene)-2,4-pentanedione ≥98.0% (by GC)
3-(1,3-Dithian-2-ylidene)-2,4-pentanedione

CAS Number: 55727-23-6
MDL Number: MFCD01096981
Molecular Formula: C9H12O2S2
Molecular Weight: 216.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103

Formula: C₉H₁₂O₂S₂
Melting Pt: 103 °C
MDL Number: MFCD01096981
CAS Number: 55727-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b7faf59fc59e5a4946c9e297f3bc92c78c69ab2&vid=0b7faf59fc59e5a4946c9e297f3bc92c78c69ab2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR