((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate ≥98.0% (by HPLC)
Supplier: TCI America
D4207-1G
D4207-5G
TCD4207-1GEA
26.86
CAD
TCD4207-1G
TCD4207-5G
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate ≥98.0% (by HPLC)
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate
CAS Number: 122111-01-7
MDL Number: MFCD08458308
Molecular Formula: C19H14F2O6
Molecular Weight: 376.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 123
Specific rotation [a]20/D: 71 deg (C=1, CH3CN)
Storage Temperature: <0°C
Formula:
C19H14F2O6 Melting Pt: 123 °C |
MDL Number:
MFCD08458308 CAS Number: 122111-01-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...