Dimethylphenylphosphine borane ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
Borane - Dimethylphenylphosphine Complex
D4204-1G
D4204-5G
TCD4204-1GEA
240.16
CAD
TCD4204-1G
TCD4204-5G
Dimethylphenylphosphine borane ≥95.0% (by HPLC)
Dimethylphenylphosphine borane
CAS Number: 35512-87-9
Molecular Formula: C8H14BP
Molecular Weight: 151.98
Purity/Analysis Method: >95.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 78
Other Product Names / Synonyms: Borane - Dimethylphenylphosphine Complex
Formula:
C₈H₁₄BP MW: 151.98 g/mol Boiling Pt: 78 °C |
CAS Number:
35512-87-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...