3,4-Dihexylthiophene ≥97.0%

Supplier: TCI America

D4202-1G D4202-5G
TCD4202-1GEA 105.86 CAD
TCD4202-1G TCD4202-5G
3,4-Dihexylthiophene ≥97.0%
3,4-Dihexylthiophene

CAS Number: 122107-04-4
MDL Number: MFCD21363049
Molecular Formula: C16H28S
Molecular Weight: 252.46
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 110
Specific Gravity (20/20): 0.91

Formula: C₁₆H₂₈S
Boiling Pt: 103 °C
Density: 0.91 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD21363049
CAS Number: 122107-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68db8263aa01b785aa2373330d47eb60f93310d0&vid=68db8263aa01b785aa2373330d47eb60f93310d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR