2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97.0%

Supplier: TCI America

Synonyms: 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester

D4201-1G D4201-5G
TCD4201-1GEA 311.26 CAD
TCD4201-1G TCD4201-5G
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97.0%
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

CAS Number: 1003298-87-0
MDL Number: MFCD20526385
Molecular Formula: C12H15BCl2O3
Molecular Weight: 288.96
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 103
Melting point (°C): 110


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester, 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Formula: C₁₂H₁₅BCl₂O₃
MW: 288.96 g/mol
Boiling Pt: 103 °C
Melting Pt: 110 °C
MDL Number: MFCD20526385
CAS Number: 1003298-87-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6170869119a228b7ade5d4976641b56f1c5eb20e&vid=6170869119a228b7ade5d4976641b56f1c5eb20e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR