3,5-Diphenyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
D4197-25G
D4197-5G
TCD4197-25GEA
432.92
CAD
TCD4197-25G
TCD4197-5G
3,5-Diphenyl-1H-pyrazole ≥98.0% (by GC)
3,5-Diphenyl-1H-pyrazole
CAS Number: 1145-01-3
MDL Number: MFCD00039675
Molecular Formula: C15H12N2
Molecular Weight: 220.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 202
Formula:
C₁₅H₁₂N₂ MW: 220.27 g/mol Melting Pt: 202 °C |
MDL Number:
MFCD00039675 CAS Number: 1145-01-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...