Phenylboronic acid neopentyl glycol ester ≥98.0%

Supplier: TCI America

Synonyms: 5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane

D4195-25G D4195-5G
TCD4195-25GEA 513.5 CAD
TCD4195-25G TCD4195-5G
Phenylboronic acid neopentyl glycol ester ≥98.0%
Phenylboronic acid neopentyl glycol ester

CAS Number: 5123-13-7
MDL Number: MFCD05663851
Molecular Formula: C11H15BO2
Molecular Weight: 190.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 65
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzene, Phenylboronic Acid Neopentyl Glycol Ester

Formula: C₁₁H₁₅BO₂
MW: 190.05 g/mol
Melting Pt: 65 °C
MDL Number: MFCD05663851
CAS Number: 5123-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b87edb1eb7446e2d1d4c26cff722e692949a666&vid=1b87edb1eb7446e2d1d4c26cff722e692949a666&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR