3-(Dimethylcarbamoyl)phenylboronic acid 97.0-111.0% (contains varying amounts of Anhydride)
Supplier: TCI America
D4194-1G
D4194-5G
TCD4194-1GEA
91.64
CAD
TCD4194-1G
TCD4194-5G
3-(Dimethylcarbamoyl)phenylboronic acid 97.0-111.0% (contains varying amounts of Anhydride)
3-(Dimethylcarbamoyl)phenylboronic acid
CAS Number: 373384-14-6
MDL Number: MFCD03412066
Molecular Formula: C9H12BNO3
Molecular Weight: 193.01
Form: Crystal
Color: White
Other Product Names / Synonyms: 3-(Dimethylcarbamoyl)benzeneboronic Acid
Formula:
C₉H₁₂BNO₃ |
MDL Number:
MFCD03412066 CAS Number: 373384-14-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...