4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥97.0%

Supplier: TCI America

Synonyms: 4-(Dimethylamino)-N-(7-(hydroxyamino)-7-oxoheptyl)benzamide, M344, M 344

D4188-100MG D4188-20MG
TCD4188-100MGEA 1294.18 CAD
TCD4188-100MG TCD4188-20MG
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥97.0%
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide

CAS Number: 251456-60-7
MDL Number: MFCD03453554
Molecular Formula: C16H25N3O3
Molecular Weight: 307.39
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 152
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antitumor Ingredients, Epigenetics Research Reagents
Other Product Names / Synonyms: M 344

Formula: C₁₆H₂₅N₃O₃
MW: 307.39 g/mol
Melting Pt: 152 °C
MDL Number: MFCD03453554
CAS Number: 251456-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e58e8754190283de430765d79be0108cb9573cc&vid=7e58e8754190283de430765d79be0108cb9573cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR