1,3-Dibromo-7-tert-butylpyrene ≥97.0% (by GC)

Supplier: TCI America

D4187-1G D4187-5G
TCD4187-1GEA 142.2 CAD
TCD4187-1G TCD4187-5G
1,3-Dibromo-7-tert-butylpyrene ≥97.0% (by GC)
1,3-Dibromo-7-tert-butylpyrene

CAS Number: 1005771-04-9
Molecular Formula: C20H16Br2
Molecular Weight: 416.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal

Formula: C₂₀H₁₆Br₂
MW: 416.16 g/mol
MDL Number: MFCD22581440
CAS Number: 1005771-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c837b0354f31d217acf1699a30aeb675b661ca56&vid=c837b0354f31d217acf1699a30aeb675b661ca56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR