2,9-Dichloro-1,10-phenanthroline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D4186-1G D4186-200MG
TCD4186-1GEA 378.57 CAD
TCD4186-1G TCD4186-200MG
2,9-Dichloro-1,10-phenanthroline ≥97.0% (by GC, titration analysis)
2,9-Dichloro-1,10-phenanthroline

CAS Number: 29176-55-4
MDL Number: MFCD02084186
Molecular Formula: C12H6Cl2N2
Molecular Weight: 249.09
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 235

Formula: C₁₂H₆Cl₂N₂
MW: 249.09 g/mol
Melting Pt: 235 °C
MDL Number: MFCD02084186
CAS Number: 29176-55-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e154fcffad028e1013f31dc06bf467ee047dcc25&vid=e154fcffad028e1013f31dc06bf467ee047dcc25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR