2,7-Di-tert-butylpyrene ≥98.0% (by GC)

Supplier: TCI America

D4185-5G D4185-1G
TCD4185-5GEA 423.44 CAD
TCD4185-5G 76219-030
2,7-Di-tert-butylpyrene ≥98.0% (by GC)
2,7-Di-tert-butylpyrene

CAS Number: 24300-91-2
Molecular Formula: C24H26
Molecular Weight: 314.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 209

Formula: C₂₄H₂₆
MW: 314.47 g/mol
Melting Pt: 209 °C
MDL Number: MFCD01013641
CAS Number: 24300-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59f77c3afb5f8768911d1153b1f698baaea75f84&vid=59f77c3afb5f8768911d1153b1f698baaea75f84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR