5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene ≥97.0% (by GC)

Supplier: TCI America

D4183-1G D4183-5G
TCD4183-1GEA 905.34 CAD
TCD4183-1G TCD4183-5G
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene ≥97.0% (by GC)
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene

CAS Number: 170702-05-3
Molecular Formula: C20H28Br2S2
Molecular Weight: 492.37
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.34
Storage Temperature: 0-10°C

Formula: C₂₀H₂₈Br₂S₂
Density: 1.34 (20 °C)
MDL Number: MFCD23380207
CAS Number: 170702-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a6adec8f9ed154daa0dfd3089ab44fcdc10175&vid=83a6adec8f9ed154daa0dfd3089ab44fcdc10175&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR