4,4'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)

Supplier: TCI America

D4181-1G D4181-5G
TCD4181-1GEA 338.12 CAD
TCD4181-1G TCD4181-5G
4,4'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)
4,4'-Dibromo-2,2'-bithiophene

CAS Number: 51285-60-0
MDL Number: MFCD00508561
Molecular Formula: C8H4Br2S2
Molecular Weight: 324.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 134

Formula: C₈H₄Br₂S₂
Melting Pt: 134 °C
MDL Number: MFCD00508561
CAS Number: 51285-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db23b12e3b38b5f69b727fb20a3fb95269d3bff0&vid=db23b12e3b38b5f69b727fb20a3fb95269d3bff0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR