3,3'-Dihexyl-2,2'-bithiophene ≥98.0% (by GC)
Supplier: TCI America
D4180-1G
D4180-5G
TCD4180-1GEA
334.96
CAD
TCD4180-1G
TCD4180-5G
3,3'-Dihexyl-2,2'-bithiophene ≥98.0% (by GC)
3,3'-Dihexyl-2,2'-bithiophene
CAS Number: 125607-30-9
Molecular Formula: C20H30S2
Molecular Weight: 334.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Specific Gravity (20/20): 1.00
Storage Temperature: 0-10°C
Formula:
C₂₀H₃₀S₂ Boiling Pt: 82 °C Density: 1.00 (20 °C) |
CAS Number:
125607-30-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...