3,5-Dibromoanthranilic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-3,5-dibromobenzoicacid, 2-Amino-3,5-dibromobenzoic acid

D4177-25G D4177-5G
TCD4177-25GEA 128.93 CAD
TCD4177-25G TCD4177-5G
3,5-Dibromoanthranilic acid ≥98.0%
3,5-Dibromoanthranilic acid

CAS Number: 609-85-8
MDL Number: MFCD00017087
Molecular Formula: C7H5Br2NO2
Molecular Weight: 294.93
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 236


Other Product Names / Synonyms: 2-Amino-3,5-dibromobenzoic Acid

Formula: C₇H₅Br₂NO₂
MW: 294.93
Melting Pt: 236 °C
MDL Number: MFCD00017087
CAS Number: 609-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd00f1156a04cc0fc379bbc7e5e92697e323d72d&vid=b10659d9b450bed8617583290394da4b8a4c09b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR