4-Decyloxybenzoic acid ≥98.0%

Supplier: TCI America

D4176-25G D4176-5G
TCD4176-25GEA 271.76 CAD
TCD4176-25G TCD4176-5G
4-Decyloxybenzoic acid ≥98.0%
4-Decyloxybenzoic acid

CAS Number: 5519-23-3
MDL Number: MFCD00020360
Molecular Formula: C17H26O3
Molecular Weight: 278.39
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 96

Formula: C₁₇H₂₆O₃
MW: 278.39 g/mol
Melting Pt: 96 °C
MDL Number: MFCD00020360
CAS Number: 5519-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad9a0b35ddbfd4da3b2c7b4d70aef9444613bb25&vid=ad9a0b35ddbfd4da3b2c7b4d70aef9444613bb25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR