trans-3,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D4161-5G
D4161-1G
TCD4161-5GEA
51.19
CAD
TCD4161-5G
TCD4161-1G
trans-3,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
trans-3,4-Difluorocinnamic acid
CAS Number: 112897-97-9
MDL Number: MFCD00010320
Molecular Formula: C9H6F2O2
Molecular Weight: 184.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 196
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: trans-3-(3,4-Difluorophenyl)acrylic Acid
Formula:
C₉H₆F₂O₂ Melting Pt: 196 °C |
MDL Number:
MFCD00010320 CAS Number: 112897-97-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...