Etidronate disodium hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dihydrogen (1-hydroxyethylidene)bisphosphonate disodium hydrate

D4159-25G D4159-5G
TCD4159-25GEA 798.37 CAD
TCD4159-25G TCD4159-5G
Etidronate disodium hydrate ≥98.0% (by titrimetric analysis)
Etidronate Disodium hydrate

CAS Number: 7414-83-7
MDL Number: MFCD00152567
Molecular Formula: C2H8O7P2
Molecular Weight: 249.99
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: Disodium 1-Hydroxyethane-1,1-diphosphonate, 1-Hydroxyethane-1,1-diphosphonic Acid Disodium Salt, Etidronic Acid Disodium Salt

Formula: C₂H₆Na₂O₇P₂·H₂O
MW: 249.99 g/mol (anhy)
MDL Number: MFCD00152567
CAS Number: 7414-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e761632b19d1153069f2d5fd5cdfce2a22dbc30a&vid=e5264051bdb6d234170d1a3f2f8e8455ddfd6bb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR