Dibenzothiophene-5,5-dioxide 98,0

Supplier: TCI America

Synonyms: Dibenzothiophene Sulphone, Dibenzosulfolane, Dibenzothiophene dioxide, Dibenzosulpholane, Dibenzothiophene sulfone, Dibenzo[b,d]thiophene 5,5-dioxide

D4153-25G D4153-5G
TCD4153-25GEA 330.22 CAD
TCD4153-25G TCD4153-5G
Dibenzothiophene-5,5-dioxide 98,0
Dibenzothiophene-5,5-dioxide

CAS Number: 1016-05-3
MDL Number: MFCD00004970
Molecular Formula: C12H8O2S
Molecular Weight: 216.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 236


Product Categories: Synthetic Organic Chemistry, Sulfones, Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Dibenzothiophene Sulfone, Dibenzosulfolane

Formula: C₁₂H₈O₂S
MW: 216.25 g/mol
Melting Pt: 236 °C
MDL Number: MFCD00004970
CAS Number: 1016-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84fff7a80660e8de900399541d1c443aa4aa9bcf&vid=84fff7a80660e8de900399541d1c443aa4aa9bcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR