1,8-Diiodoanthracene ≥98.0% (by GC)

Supplier: TCI America

D4151-1G D4151-5G
TCD4151-1GEA 265.44 CAD
TCD4151-1G TCD4151-5G
1,8-Diiodoanthracene ≥98.0% (by GC)
1,8-Diiodoanthracene

CAS Number: 189105-78-0
Molecular Formula: C14H8I2
Molecular Weight: 430.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 201
Storage Temperature: <0°C

Formula: C₁₄H₈I₂
Melting Pt: 201 °C
CAS Number: 189105-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60678e38c35a2ad8e54d8ae9147160b1b48f72fd&vid=60678e38c35a2ad8e54d8ae9147160b1b48f72fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR