10-Deacetylbaccatin III ≥98.0% (by HPLC)

Supplier: TCI America

D4148-100MG D4148-500MG
TCD4148-100MGEA 165.27 CAD
TCD4148-100MG TCD4148-500MG
10-Deacetylbaccatin III ≥98.0% (by HPLC)
10-Deacetylbaccatin III

CAS Number: 32981-86-5
MDL Number: MFCD00132913
Molecular Formula: C29H36O10
Molecular Weight: 544.60
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 234
Specific rotation [a]20/D: -42 deg (C=1, MeOH)
Storage Temperature: 0-10°C

Formula: C₂₉H₃₆O₁₀
Melting Pt: 234 °C
MDL Number: MFCD00132913
CAS Number: 32981-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=221a025f8048db345b01bb49a70878a838c51d08&vid=221a025f8048db345b01bb49a70878a838c51d08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR