2-((Diphenylmethylene)amino)acetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-(Diphenylmethylene)aminoacetonitrile

D4145-25G D4145-5G
TCD4145-25GEA 413.96 CAD
TCD4145-25G TCD4145-5G
2-((Diphenylmethylene)amino)acetonitrile ≥98.0% (by GC)
2-((Diphenylmethylene)amino)acetonitrile

CAS Number: 70591-20-7
MDL Number: MFCD00009970
Molecular Formula: C15H12N2
Molecular Weight: 220.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85
Lambda max.: 245 nm (H2O)Storage Temperature: 0-10°C

Formula: C₁₅H₁₂N₂
MW: 220.2700 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00009970
CAS Number: 70591-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7acf9f38ac26f86a556f16699177749d8b85f7b&vid=a7acf9f38ac26f86a556f16699177749d8b85f7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR