3,5-Dinitrobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

D4144-25G D4144-5G
TCD4144-25GEA 415.54 CAD
TCD4144-25G TCD4144-5G
3,5-Dinitrobenzyl alcohol ≥98.0% (by GC)
3,5-Dinitrobenzyl alcohol

CAS Number: 71022-43-0
MDL Number: MFCD00007235
Molecular Formula: C7H6N2O5
Molecular Weight: 198.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91

Formula: C₇H₆N₂O₅
Melting Pt: 91 °C
MDL Number: MFCD00007235
CAS Number: 71022-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a2d4ed8284a26a4855131e0e965d85f1a03155c&vid=4a2d4ed8284a26a4855131e0e965d85f1a03155c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR