4,6-Dichloro-2-(methylsulfonyl)pyrimidine ≥98.0%

Supplier: TCI America

Synonyms: 4,6-Dichloro-2-methylsulfonylpyrimidine, 4,6-Dichloro-2-(methanesulphonyl)pyrimidine, 4,6-Dichloro-2-methylsulphonylpyrimidine, 4,6-Dichloro-2-(methanesulfonyl)pyrimidine

D4135-1G D4135-5G
TCD4135-1GEA 47.4 CAD
TCD4135-1G TCD4135-5G
4,6-Dichloro-2-(methylsulfonyl)pyrimidine ≥98.0%
4,6-Dichloro-2-(methylsulfonyl)pyrimidine

CAS Number: 4489-34-3
MDL Number: MFCD00672147
Molecular Formula: C5H4Cl2N2O2S
Molecular Weight: 227.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 120


Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: 4,6-Dichloro-2-(methanesulfonyl)pyrimidine

Formula: C₅H₄Cl₂N₂O₂S
MW: 227.06 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00672147
CAS Number: 4489-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fb09d0474c8a76c1fba13ac08b0e45a804f7189&vid=2fb09d0474c8a76c1fba13ac08b0e45a804f7189&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR