(S)-O-Desmethylraclopride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4134-1G D4134-200MG
TCD4134-1GEA 484.27 CAD
TCD4134-1G TCD4134-200MG
(S)-O-Desmethylraclopride ≥98.0% (by HPLC, titration analysis)
(S)-O-Desmethylraclopride

CAS Number: 119670-11-0
MDL Number: MFCD02179385
Molecular Formula: C14H18Cl2N2O3
Molecular Weight: 333.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 223
Specific rotation [a]20/D: -31 deg (C=1, DMSO)


Other Product Names / Synonyms: (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide

Formula: C₁₄H₁₈Cl₂N₂O₃
Melting Pt: 223 °C
MDL Number: MFCD02179385
CAS Number: 119670-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c308caf01ba0afb5f0c57c52716e555a42c6e5c&vid=7c308caf01ba0afb5f0c57c52716e555a42c6e5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR