3,5-Di-tert-butylbenzaldehyde ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 3,5-Bis(tert-butyl)benzaldehyde

D4130-1G D4130-5G
TCD4130-1GEA 71.1 CAD
TCD4130-1G TCD4130-5G
3,5-Di-tert-butylbenzaldehyde ≥95.0% (by GC)
3,5-Di-tert-butylbenzaldehyde

CAS Number: 17610-00-3
MDL Number: MFCD00026298
Molecular Formula: C15H22O
Molecular Weight: 218.34
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 86
Lambda max.: 296 nm (CHCl3)

Formula: C₁₅H₂₂O
MW: 218.34
Boiling Pt: 132 °C
Melting Pt: 86 °C
MDL Number: MFCD00026298
CAS Number: 17610-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3595e5c0e61b65fb3b496706ba593464cb6bfb8&vid=f3595e5c0e61b65fb3b496706ba593464cb6bfb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR