1,3-Difluoro-2-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

D4123-1G D4123-5G
TCD4123-1GEA 79 CAD
TCD4123-1G TCD4123-5G
1,3-Difluoro-2-nitrobenzene ≥98.0% (by GC)
1,3-Difluoro-2-nitrobenzene

CAS Number: 19064-24-5
MDL Number: MFCD00192035
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 92
Flash Point (°C): 88
Freezing point (°C): 22


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3-Difluoro-2-nitrobenzene

Formula: C₆H₃F₂NO₂
Boiling Pt: 92 °C
Flash Pt: 88 °C
MDL Number: MFCD00192035
CAS Number: 19064-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=164ad9030613a7ef7d52b9e2d407fb8d024de7bb&vid=164ad9030613a7ef7d52b9e2d407fb8d024de7bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR