2,4-Dichloro-5-iodopyrimidine ≥98.0% (by GC)

Supplier: TCI America

D4122-1G D4122-5G
TCD4122-1GEA 67.94 CAD
TCD4122-1G TCD4122-5G
2,4-Dichloro-5-iodopyrimidine ≥98.0% (by GC)
2,4-Dichloro-5-iodopyrimidine

CAS Number: 13544-44-0
MDL Number: MFCD01898087
Molecular Formula: C4HCl2IN2
Molecular Weight: 274.87
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₄HCl₂IN₂
MW: 274.87 g/mol
Melting Pt: 70 °C
MDL Number: MFCD01898087
CAS Number: 13544-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=452d5f8299046a83d34e9d21e5573e327b0f7f08&vid=452d5f8299046a83d34e9d21e5573e327b0f7f08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR