2',6'-Dichloroacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(2,6-Dichlorophenyl)ethanone
D4119-1G
D4119-5G
TCD4119-1GEA
50.56
CAD
TCD4119-1G
TCD4119-5G
2',6'-Dichloroacetophenone ≥98.0%
2',6'-Dichloroacetophenone
CAS Number: 2040-05-3
MDL Number: MFCD00016339
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 141
Melting point (°C): 44
Formula:
C₈H₆Cl₂O MW: 189.04 Boiling Pt: 141 °C Melting Pt: 44 °C |
MDL Number:
MFCD00016339 CAS Number: 2040-05-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...