3,5-Dibromo-4-picoline ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dibromo-4-methylpyridine

D4118-1G D4118-5G
TCD4118-1GEA 63.2 CAD
TCD4118-1G TCD4118-5G
3,5-Dibromo-4-picoline ≥98.0%
3,5-Dibromo-4-picoline

CAS Number: 3430-23-7
MDL Number: MFCD00128859
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: 3,5-Dibromo-4-picoline

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00128859
CAS Number: 3430-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a0d9cdbb243421b9784efbd3aaf07a4f8bd095&vid=b7a0d9cdbb243421b9784efbd3aaf07a4f8bd095&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR