trans-2,6-Difluorocinnamic acid ≥95.0%
Supplier: TCI America
Synonyms:
(E)-3-(2,6-Difluorophenyl)acrylic acid
D4115-1G
D4115-5G
TCD4115-1GEA
60.04
CAD
TCD4115-1G
TCD4115-5G
trans-2,6-Difluorocinnamic acid ≥95.0%
trans-2,6-Difluorocinnamic acid
CAS Number: 102082-89-3
MDL Number: MFCD00010319
Molecular Formula: C9H6F2O2
Molecular Weight: 184.14
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 122
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: trans-3-(2,6-Difluorophenyl)acrylic Acid
Formula:
C₉H₆F₂O₂ Melting Pt: 122 °C |
MDL Number:
MFCD00010319 CAS Number: 102082-89-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...