Di-tert-butyl Iminodiacetate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Iminodiacetic acid Di-tert-butyl Ester, Di-tert-butyl 2,2'-azanediyldiacetate

D4109-1G D4109-5G
TCD4109-1GEA 91.64 CAD
TCD4109-1G TCD4109-5G
Di-tert-butyl Iminodiacetate ≥97.0% (by GC)
Di-tert-butyl Iminodiacetate

CAS Number: 85916-13-8
MDL Number: MFCD01863672
Molecular Formula: C12H23NO4
Molecular Weight: 245.32
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 42


Other Product Names / Synonyms: Iminodiacetic Acid Di-tert-butyl Ester

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
Melting Pt: 42 °C
MDL Number: MFCD01863672
CAS Number: 85916-13-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2026b633f6577990edea5d4e532596a1c6b45b1&vid=d2026b633f6577990edea5d4e532596a1c6b45b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR