Diflufenican ≥98.0%

Supplier: TCI America

Synonyms: N-(2,4-Difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]nicotinamide

D4108-25G D4108-5G
TCD4108-25GEA 630.42 CAD
TCD4108-25G TCD4108-5G
Diflufenican ≥98.0%
Diflufenican

CAS Number: 83164-33-4
MDL Number: MFCD01311804
Molecular Formula: C19H11F5N2O2
Molecular Weight: 394.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 161


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: N-(2,4-Difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]nicotinamide

Formula: C₁₉H₁₁F₅N₂O₂
MW: 394.30 g/mol
Melting Pt: 161 °C
MDL Number: MFCD01311804
CAS Number: 83164-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b6714497e838677eea82199e5a1f9816e5f28a&vid=c3b6714497e838677eea82199e5a1f9816e5f28a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR