2-(3,4-Dimethoxyphenyl)ethanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol

D4107-25G D4107-5G
TCD4107-25GEA 230.68 CAD
TCD4107-25G TCD4107-5G
2-(3,4-Dimethoxyphenyl)ethanol ≥98.0% (by GC)
2-(3,4-Dimethoxyphenyl)ethanol

CAS Number: 7417-21-2
MDL Number: MFCD00002894
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 46
Flash Point (°C): 113


Other Product Names / Synonyms: 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol, Homoveratryl Alcohol

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 174 °C
Melting Pt: 46 °C
Flash Pt: 113 °C
MDL Number: MFCD00002894
CAS Number: 7417-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8899ba7d8de0456381c68564e5fb6c51cf969b2a&vid=8899ba7d8de0456381c68564e5fb6c51cf969b2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR