4,6-Dichloro-5-methylpyrimidine ≥98.0% (by GC)

Supplier: TCI America

D4091-25G D4091-5G
TCD4091-25GEA 520.61 CAD
TCD4091-25G TCD4091-5G
4,6-Dichloro-5-methylpyrimidine ≥98.0% (by GC)
4,6-Dichloro-5-methylpyrimidine

CAS Number: 4316-97-6
MDL Number: MFCD01646112
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59

Formula: C₅H₄Cl₂N₂
Melting Pt: 59 °C
MDL Number: MFCD01646112
CAS Number: 4316-97-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5d38d5a024834c3e2b790f446908574d953cc1c&vid=b5d38d5a024834c3e2b790f446908574d953cc1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR