2,7-Di-tert-butylfluorene ≥98.0% (by GC)
Supplier: TCI America
D4084-25G
D4084-5G
TCD4084-25GEA
292.62
CAD
TCD4084-25G
TCD4084-5G
2,7-Di-tert-butylfluorene ≥98.0% (by GC)
2,7-Di-tert-butylfluorene
CAS Number: 58775-05-6
MDL Number: MFCD03093998
Molecular Formula: C21H26
Molecular Weight: 278.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124
Formula:
C₂₁H₂₆ MW: 278.43 g/mol Melting Pt: 124 °C |
MDL Number:
MFCD03093998 CAS Number: 58775-05-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...