Dimetridazole ≥97.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 1,2-Dimethyl-5-nitroimidazole

D4081-25G D4081-5G
TCD4081-25GEA 116.92 CAD
TCD4081-25G TCD4081-5G
Dimetridazole ≥97.0% (by titrimetric analysis) for biochemical research
Dimetridazole

[for Biochemical Research]
CAS Number: 551-92-8
MDL Number: MFCD00047046
Molecular Formula: C5H7N3O2
Molecular Weight: 141.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 140
Storage Temperature: 0-10°C


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Dimetridazole

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00047046
CAS Number: 551-92-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60673efb110015d64cfadee236520882b40e229e&vid=de4770366a422d5513ed09713d775612181ba815&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR