3,4-Bis(benzyloxy)benzaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
3,4-Dibenzyloxybenzaldehyde
D4079-25G
D4079-5G
TCD4079-25GEA
344.44
CAD
TCD4079-25G
TCD4079-5G
3,4-Bis(benzyloxy)benzaldehyde ≥98.0%
3,4-Bis(benzyloxy)benzaldehyde
CAS Number: 5447-02-9
MDL Number: MFCD00004776
Molecular Formula: C21H18O3
Molecular Weight: 318.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92
Other Product Names / Synonyms: 3,4-Bis(benzyloxy)benzaldehyde
Formula:
C₂₁H₁₈O₃ Melting Pt: 92 °C |
MDL Number:
MFCD00004776 CAS Number: 5447-02-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...