4-(4,6-Dichloropyrimidin-2-yl)morpholine ≥98.0% (by GC)

Supplier: TCI America

D4069-5G D4069-1G
TCD4069-5GEA 306.52 CAD
TCD4069-5G TCD4069-1G
4-(4,6-Dichloropyrimidin-2-yl)morpholine ≥98.0% (by GC)
4-(4,6-Dichloropyrimidin-2-yl)morpholine

CAS Number: 10397-13-4
MDL Number: MFCD05022359
Molecular Formula: C8H9Cl2N3O
Molecular Weight: 234.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 141


Other Product Names / Synonyms: 2-(4-Morpholino)-4,6-dichloropyrimidine

Formula: C₈H₉Cl₂N₃O
Melting Pt: 141 °C
MDL Number: MFCD05022359
CAS Number: 10397-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb1fbc28c4bdf6d8b25381892fdbb39ce56eaf48&vid=eb1fbc28c4bdf6d8b25381892fdbb39ce56eaf48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR