Dimethyl-5-bromoisophthalate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester
CAS Number: 51760-21-5
MDL Number: MFCD00078709
Molecular Formula: C10H9BrO4
Molecular Weight: 273.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 89
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 5-Bromoisophthalic Acid Dimethyl Ester
Formula:
C₁₀H₉BrO₄ MW: 273.08 g/mol Boiling Pt: 159 °C Melting Pt: 89 °C |
MDL Number:
MFCD00078709 CAS Number: 51760-21-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...