2,5-Dibromohydroquinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Dibromobenzene-1,4-diol

D4067-25G D4067-5G
TCD4067-25GEA 276.5 CAD
TCD4067-25G TCD4067-5G
2,5-Dibromohydroquinone ≥98.0% (by GC, titration analysis)
2,5-Dibromohydroquinone

CAS Number: 14753-51-6
MDL Number: MFCD00192664
Molecular Formula: C6H4Br2O2
Molecular Weight: 267.90
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 193


Other Product Names / Synonyms: 2,5-Dibromo-1,4-benzenediol

Formula: C₆H₄Br₂O₂
MW: 267.9 g/mol
Melting Pt: 193 °C
MDL Number: MFCD00192664
CAS Number: 14753-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60d07e4f378d1d7245559764083af298523777d&vid=d60d07e4f378d1d7245559764083af298523777d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR