4,5-Difluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4063-25G D4063-5G
TCD4063-25GEA 722.06 CAD
TCD4063-25G TCD4063-5G
4,5-Difluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)
4,5-Difluoroanthranilic acid

CAS Number: 83506-93-8
MDL Number: MFCD00077509
Molecular Formula: C7H5F2NO2
Molecular Weight: 173.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-4,5-difluorobenzoic Acid

Formula: C₇H₅F₂NO₂
Melting Pt: 180 °C
MDL Number: MFCD00077509
CAS Number: 83506-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cc65d5634bcb70bd8ded3a3f8cc53460b23484&vid=63cc65d5634bcb70bd8ded3a3f8cc53460b23484&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR